Skip to Content
Merck
All Photos(1)

Documents

40859

Sigma-Aldrich

1-Bromo-4-(heptadecafluorooctyl)benzene

≥95.0%

Synonym(s):

1-Bromo-4-(perfluorooctyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H4BrF17
CAS Number:
Molecular Weight:
575.06
Beilstein:
7895098
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

mp

35-40 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)c1ccc(Br)cc1

InChI

1S/C14H4BrF17/c15-6-3-1-5(2-4-6)7(16,17)8(18,19)9(20,21)10(22,23)11(24,25)12(26,27)13(28,29)14(30,31)32/h1-4H

InChI key

XDEOJAJPLXXYKA-UHFFFAOYSA-N

Other Notes

Preparation of perfluoroalkylphenyl derivatives; Synthesis of a perfluoro-tagged phosphine for Stille couplings in a fluorous biphasic system

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.J.L. Paciorek et al.
Journal of Fluorine Chemistry, 88, 55-55 (1998)
Repetitive Application of Perfluoro-Tagged Pd Complexes for Stille Couplings in a Fluorous Biphasic System This work was supported by Fluka (Buchs) and Byk Gulden Pharmaceuticals (Konstanz). S.S. thanks Fluka for financial support. We would also thank Priv.-Doz. Dr. C. Rücker for help in the preparation of the manuscript.
Schneider et al.
Angewandte Chemie (International ed. in English), 39(22), 4142-4145 (2000-11-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service