Skip to Content
Merck
All Photos(1)

Key Documents

156728

Sigma-Aldrich

6-Methyl-4-chromanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

141-143 °C/13.5 mmHg (lit.)

mp

31-35 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

Cc1ccc2OCCC(=O)c2c1

InChI

1S/C10H10O2/c1-7-2-3-10-8(6-7)9(11)4-5-12-10/h2-3,6H,4-5H2,1H3

InChI key

RJHXEPLSJAVTFW-UHFFFAOYSA-N

Application

The product has been used as a substrate for carbonyl reductase from Sporobolomyces salmonicolor, wherein it was reduced to the corresponding (R)-chiral alcohol. It has also been used as a substrate for the enantioselective reduction by 3 biocatalysts, namely, Didymosphaeria igniaria, Coryneum betulinum and Chaetomium sp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective reduction of 4-chromanone and its derivatives by selected filamentous fungi.
Janeczko T, et al.
Journal of Molecular Catalysis. B, Enzymatic, 97, 278-282 (2013)
Dunming Zhu et al.
Organic & biomolecular chemistry, 4(14), 2690-2695 (2006-07-11)
In our effort to search for effective carbonyl reductases, the activity and enantioselectivity of a carbonyl reductase from Sporobolomyces salmonicolor have been evaluated toward the reduction of a variety of ketones. This carbonyl reductase (SSCR) reduces a broad spectrum of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service