Skip to Content
Merck
All Photos(1)

Key Documents

139823

Sigma-Aldrich

4-Methyl-3-nitroanisole

99%

Synonym(s):

4-Methoxy-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OCH3
CAS Number:
Molecular Weight:
167.16
Beilstein:
2047309
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

266-267 °C (lit.)

mp

17 °C (lit.)

density

1.207 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(C)c(c1)[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-3-4-7(12-2)5-8(6)9(10)11/h3-5H,1-2H3

InChI key

JBORNNNGTJSTLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methyl-3-nitroanisole was used as external standard in synthesis of antibody catalysts by bait and switch hapten strategy. It was used as starting material for synthesis of 4-fluoro and 2-fluoro-substituted acetylcholinesterase inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Wentworth et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(11), 5971-5975 (1998-05-30)
General base catalysis supplied by the histidine-12 (H-12) residue of ribonuclease (RNase) A has long been appreciated as a major component of the catalytic power of the enzyme. In an attempt to harness the catalytic power of a general base
Sang-Yoon Lee et al.
Nuclear medicine communications, 25(6), 591-596 (2004-05-29)
Acetylcholinesterase (AChE) is an important cholinergic marker for the diagnosis of Alzheimer's disease (AD). A recent study has demonstrated that C-labelled 5,7-dihydro-7-methyl-3-[2-[1-(phenylmethyl)-4-piperidinyl]ethyl]-6H-pyrrolo[3,2-f]-1,2-benzisoxazol-6-one (CP-126,998) shows promising results. The demethylated form of this ligand (CP-118,954) is a more potent and selective inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service