Merck
  • A regulatory sub-circuit downstream of Wnt signaling controls developmental transitions in neural crest formation.
A regulatory sub-circuit downstream of Wnt signaling controls developmental transitions in neural crest formation.
PLoS genetics (2021-01-20)
Ana Paula Azambuja, Marcos Simoes-Costa
ABSTRACT
The process of cell fate commitment involves sequential changes in the gene expression profiles of embryonic progenitors. This is exemplified in the development of the neural crest, a migratory stem cell population derived from the ectoderm of vertebrate embryos. During neural crest formation, cells transition through distinct transcriptional states in a stepwise manner. The mechanisms underpinning these shifts in cell identity are still poorly understood. Here we employ enhancer analysis to identify a genetic sub-circuit that controls developmental transitions in the nascent neural crest. This sub-circuit links Wnt target genes in an incoherent feedforward loop that controls the sequential activation of genes in the neural crest lineage. By examining the cis-regulatory apparatus of Wnt effector gene AXUD1, we found that multipotency factor SP5 directly promotes neural plate border identity, while inhibiting premature expression of specification genes. Our results highlight the importance of repressive interactions in the neural crest gene regulatory network and illustrate how genes activated by the same upstream signal become temporally segregated during progressive fate restriction.
Sigma-Aldrich
ChIPAb+ LEF1 - ChIP Validated Antibody and Primer Set, from mouse