SAAR3063040
Hydrochloric acid
32%, univAR™
Synonym(s):
Hydrochloric acid
About This Item
Recommended Products
product name
Hydrochloric acid 32%, univAR™
vapor density
1.3 (vs air)
vapor pressure
3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)
product line
univAR™
packaging
glass bottle of 4 × 2.5 L (SAAR3063040LC)
poly bottle of 4 × 2.5 L (SAAR3063040LP)
poly bottle of 4 × 5 L (SAAR3063040LL)
bp
>100 °C (lit.)
density
1.070 g/cm3 at 20 °C (lit.)
storage temp.
2-30°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service