Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C17H11F3N2O2S
CAS Number:
Molecular Weight:
364.34
MDL number:
NACRES:
NA.28
Assay:
≥95% (HPLC)
Form:
powder
Quality Segment
assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(C1=CC=C(NC2=NC(C(F)(F)F)=C(C3=CC=CC=C3)S2)C=C1)O
Biochem/physiol Actions
Selective Orai1 and SOCE (store-operated Ca2+ entry) potentiator that marginally affects Orai2 and Orai3.
Orai1 Agonist IA65 is a selective Orai1 and SOCE (store-operated Ca2+ entry) potentiator that preferentially occupies Oari1 N-terminal domain binding pocket and significantly enhances Orai1-mediated Ca2+-influx and concurrently potentiates Orai1 Ca2+-dependent inactivation (CDI) with an EC50 of 1.9 uM. IA65 mildly potentiates ORAI3-mediated ICRAC with a slight inhibition of ICRAC mediated by ORAI2. Increases Orai1 activity in VSMCs and skeletal muscle fibers in a concentration-dependent manner. IA-65 is shown to be inactive against TRPV1, TRPM8, or CaV2.2 and nontoxic up to 100 uM.
Orai1 Agonist IA65 is a selective Orai1 and SOCE (store-operated Ca2+ entry) potentiator that preferentially occupies Oari1 N-terminal domain binding pocket and significantly enhances Orai1-mediated Ca2+-influx and concurrently potentiates Orai1 Ca2+-dependent inactivation (CDI) with an EC50 of 1.9 uM. IA65 mildly potentiates ORAI3-mediated ICRAC with a slight inhibition of ICRAC mediated by ORAI2. Increases Orai1 activity in VSMCs and skeletal muscle fibers in a concentration-dependent manner. IA-65 is shown to be inactive against TRPV1, TRPM8, or CaV2.2 and nontoxic up to 100 uM.
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
