MTCP411311
MTC™ Bio Transfer Pipette
size 5 mL, large bulb, sterile, pack of 500 ea (individually wrapped)
Synonym(s):
pipet
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
low-density polyethylene
sterility
sterile
feature
graduation
packaging
pack of 500 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio P4113-11
L
160 mm
size
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Transfer pipette, 5ml, large bulb, graduated to 1mL, sterile, individually wrapped, 500/cs
Features and Benefits
- Transparent, non-toxic and inert
- Precise, uniform drop size
- Shatter-proof and unbreakable
- Heat Sealable
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service