Skip to Content
Merck
All Photos(1)

Key Documents

264150

Sigma-Aldrich

Caspase-3 Substrate IV, Fluorogenic

Fluorogenic caspase-3 substrate.

Synonym(s):

Caspase-3 Substrate IV, Fluorogenic, Z-DEVD-AFC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H38F3N5O14
CAS Number:
Molecular Weight:
821.71
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

DMSO: 10 mg/mL

fluorescence

λex ~400 nm
λem ~505 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

FC(F)(F)C1=CC(=O)Oc2c1ccc(c2)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)OCc3ccccc3)CC(=O)OC)CCC(=O)OC)C(C)C)CC(=O)OC

InChI key

NLDGPABVDARZCY-KIAQLKRMSA-N

General description

Fluorogenic CPP-32/apopain substrate. Also acts as a substrate for granzyme B-processed ICE-LAP3. Reaction can be monitored by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Excitation max: ~400 nm; emission max: ~505 nm.
Fluorogenic caspase-3 substrate. Also acts as a substrate for granzyme B-processed ICE-LAP3. Reaction can be monitored by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Also a substrate for caspase-6, caspase-7, caspase-8, and caspase-10.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Fluorogenic caspase-3 substrate.
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Asp-Glu-Val-Asp-AFC

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 3 months at -20°C.

Other Notes

Keramaris, E., et al. 2000. Mol. Cell Neurosci. 15, 368.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Kluck, R.M., et al. 1997. Science275, 1132.
Talanian, J., et al. 1997. J. Biol. Chem.272, 9677.
Chinnaiyan, A.M., et al. 1996. Curr. Biol.6, 897.
Nicholson, D.W., et al. 1995. Nature376, 37.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service