Key Documents
MT51340034
Mettler Toledo Electrolyte for ISE
For Nitrate ISE, volume 20 mL
About This Item
UNSPSC Code:
51191600
NACRES:
NB.79
form
liquid
manufacturer/tradename
Mettler-Toledo 51340034
volume
20 mL
General description
Achive top precision with the right electrolyte. METTLER TOLEDO offers electrolytes for every ISE so fill-up your electrode with correct and fresh electrolyte for a precise ion determination.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)