Merck
Dapsone and sulfapyridine.
Dermatologic clinics (2001-01-13)
U Paniker, N Levine
ABSTRACT
Dapsone and sulfapyridine are structurally related compounds with anti-microbial and anti-inflammatory effects. Dapsone remains the most important drug for leprosy and is useful in the prophylaxis of Pneumocystis pneumonia in patients with HIV disease. The medical treatment of choice for dermatitis herpetiformis is dapsone; and sulfapyridine also can be used for those patients who are intolerant of dapsone. Other neutrophilic disorders also may respond to these drugs. Toxic side effects of both dapsone and sulfapyridine are mediated through the hydroxylamine metabolite. These include hemolysis, methemoglobinemia, and agranulocytosis. Careful monitoring for possible adverse reactions includes frequently performing complete blood counts and regular blood chemistry profile determinations.
Product Description
Supelco
Sulfapyridine melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfapyridine, VETRANAL®, analytical standard
Supelco
Sulfapyridine, ≥99.0%