Merck
  • The Activation of ROS/NF-κB/MMP-9 Pathway Promotes Calcium-Induced Kidney Crystal Deposition.
The Activation of ROS/NF-κB/MMP-9 Pathway Promotes Calcium-Induced Kidney Crystal Deposition.
Oxidative medicine and cellular longevity (2021-07-03)
Yue Wu, Jiaqiao Zhang, Cong Li, Henglong Hu, Baolong Qin, Tao Wang, Yuchao Lu, Shaogang Wang
ABSTRACT
Idiopathic hypercalciuria is an important risk factor for the formation of calcium-containing kidney stones. Matrix metalloproteinase-9 (MMP-9) is closely related to cell and tissue remodeling and is involved in ectopic tissue calcification. However, little is known about its role in kidney stone formation. In this study, we found that the expression of MMP-9 and that of osteoblastic-related proteins was increased in normal rat kidney epithelial-like (NRK-52E) cells following treatment with a high concentration of calcium, while the knockout or overexpression of MMP-9 could, respectively, significantly inhibit or upregulate the expression of osteoblastic-related proteins and calcium crystal deposition. In addition, apoptosis and calcium crystal deposition were significantly reduced in Sprague-Dawley rats with 1,25(OH)2D3-induced hypercalciuria following MMP-9 inhibitor I treatment. Furthermore, inhibiting reactive oxygen species (ROS) production or the nuclear factor kappa-light-chain-enhancer of activated B cell (NF-κB) pathway significantly reduced calcium-induced MMP-9 expression and calcium crystal deposition. In summary, our results suggested that a high calcium concentration promotes epithelial-osteoblastic transformation and calcium crystal deposition in renal tubule cells by regulating the ROS/NF-κB/MMP-9 axis and identified a novel role for MMP-9 in regulating calcium-induced calcium crystal deposition in renal tubules.
Sigma-Aldrich
Decamethylcyclopentasiloxane, 97%
Sigma-Aldrich
Bovine Serum Albumin, ≥98%, ≤5% Loss on drying, suitable for western blot