V900933
Bovine Serum Albumin
≥98%, ≤5% Loss on drying, suitable for western blot
Synonym(s):
Bovine Serum Albumin, Albumin bovine serum, BSA, Bovine albumin
Select a Size
Select a Size
About This Item
Recommended Products
biological source
bovine
Quality Level
grade
reagent grade
product line
Vetec™
Assay
≥98%
form
lyophilized powder
mol wt
~66 kDa
~66 kDa
technique(s)
western blot: suitable
impurities
≤5% Loss on drying
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Preparation Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service