Merck
  • Silica nanoparticles with Tb(III)-centered luminescence decorated by Ag0 as efficient cellular contrast agent with anticancer effect.
Silica nanoparticles with Tb(III)-centered luminescence decorated by Ag0 as efficient cellular contrast agent with anticancer effect.
Journal of inorganic biochemistry (2018-02-28)
Svetlana V Fedorenko, Svetlana L Grechkina, Alsu R Mukhametshina, Anastasiya O Solovieva, Tatiana N Pozmogova, Svetlana M Miroshnichenko, Alexander Y Alekseev, Michael A Shestopalov, Kirill V Kholin, Irek R Nizameev, Asiya R Mustafina
ABSTRACT
The present work introduces composite luminescent nanoparticles (Ag0-Tb3+-SNs), where ultra-small nanosilver (4 ± 2 nm) is deposited onto amino-modified silica nanoparticles (35±6 nm) doped by green luminescent Tb(III) complexes. Ag0-Tb3+-SNs are able to image cancer (Hep-2) cells in confocal microscopy measurements due to efficient cell internalization, which is confirmed by TEM images of the Hep-2 cells exposed by Ag0-Tb3+-SNs. Comparative analysis of the cytotoxicity of normal fibroblasts (DK-4) and cancer cells (Hep-2) incubated with various concentrations of Ag0-Tb3+-SNs revealed the concentration range where the toxic effect on the cancer cells is significant, while it is insignificant towards the nonmalignant fibroblasts cells. The obtained results reveal Ag0-Tb3+-SNs as good cellular contrast agent able to induce the cancer cells death, which makes them promising theranostic in cancer diagnostics and therapy.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Terbium(III) nitrate hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, AR, ≥99%