Skip to Content
Merck
All Photos(2)

Key Documents

V900919

Sigma-Aldrich

Amphotericin B from Streptomyces sp.

suitable for cell culture, Vetec, BioReagent

Synonym(s):

Amphotericin B from Streptomyces sp., Fungizone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Beilstein:
78342
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

Amphotericin B from Streptomyces sp., Vetec, reagent grade, BioReagent, suitable for cell culture, ~80%

grade

reagent grade

Quality Level

product line

BioReagent
Vetec

Assay

~80%

technique(s)

cell culture | mammalian: suitable

antibiotic activity spectrum

fungi
yeast

Mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@@H](O[C@@H]2C[C@@H]3O[C@](O)(C[C@@H](O)C[C@@H](O)[C@H](O)CC[C@@H](O)C[C@@H](O)CC(=O)O[C@@H](C)[C@H](C)[C@H](O)[C@@H](C)\C=C\C=C\C=C\C=C\C=C\C=C\C=C\2)C[C@H](O)[C@H]3C(O)=O)[C@@H](O)[C@@H](N)[C@@H]1O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5+,9-7+,10-8+,13-11+,14-12+,17-15+,18-16+/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-INPOYWNPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Amphotericin B binds to sterols, forming pores in the membrane, and causing small molecules to leak out. The product is most effective against fungi and yeast.
Amphotericin B is used for primary treatment of acute invasive fungal infections, such as aspergillosis.[1][2]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Voriconazole versus amphotericin B for primary therapy of invasive aspergillosis.
Herbrecht R, et al.
The New England Journal of Medicine, 347(6), 408-415 (2002)
Randomized, double-blind clinical trial of amphotericin B colloidal dispersion vs. amphotericin B in the empirical treatment of fever and neutropenia.
White MH, et al.
Clinical Infectious Diseases, 27(2), 296-302 (1998)
Chitralekha Mohanty et al.
Anticancer research, 34(6), 2801-2806 (2014-06-13)
The use of ion radiation therapy is growing due to the continuously increasing positive clinical experience obtained. Therefore, there is a high interest in radio-biological experiments comparing the relative efficiency in cell killing of ions and photons as photons are
Malav S Trivedi et al.
The Journal of nutritional biochemistry, 25(10), 1011-1018 (2014-07-16)
Dietary interventions like gluten-free and casein-free diets have been reported to improve intestinal, autoimmune and neurological symptoms in patients with a variety of conditions; however, the underlying mechanism of benefit for such diets remains unclear. Epigenetic programming, including CpG methylation
Rujiu Hu et al.
Journal of animal science and biotechnology, 12(1), 25-25 (2021-02-18)
Lactobacillus reuteri strains are widely used as probiotics to prevent and treat inflammatory bowel disease by modulating the host's immune system. However, the underlying mechanisms by which they communicate with the host have not been clearly understood. Bacterial extracellular vesicles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service