Skip to Content
Merck
All Photos(2)

Key Documents

V900863

Sigma-Aldrich

Porcine Gelatin

from porcine skin, Type A, powder, gel strength ~300 g Bloom, Vetec

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202

Product Name

Gelatin from porcine skin, Vetec, reagent grade, Type A, powder, gel strength ~300 g Bloom

biological source

Porcine skin

grade

reagent grade

type

Type A

product line

Vetec

form

powder

packaging

poly bottle of 100 g
poly bottle of 500 g

dilution

(biocompatible polymer: suitable)
(coating cell culture: suitable)

solubility

H2O: soluble 50 mg/mL, clear to hazy, faintly yellow to yellow

Looking for similar products? Visit Product Comparison Guide

General description

Gelatin acts as a key element in a variety of foodstuffs and pharmaceutical products, including jellies, aspics, desserts, capsule, tablets and blood substitutes. Usually, most gelatin products have been manufactured from porcine skin.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juyi Wan et al.
Artificial organs, 45(9), 1068-1082 (2021-03-18)
To prepare a tissue-engineered pulmonary valved conduit (PVC) with good tensile strength and biocompatibility. Sixty adult porcine PVCs were used to determine the optimal decellularization time. Five juvenile porcine decellularized PVCs and five juvenile porcine crosslinked PVCs were subsequently prepared
Xing Qiu et al.
Journal of anatomy, 229(2), 326-333 (2016-04-15)
We describe a technique for perfusing a barium sulphate suspension into the intraosseous artery. Following the perfusion of abarium sulphate suspension into 14 fresh lower limbs of Chinese cadavers, micro-CT scanning was applied to digitize, quantify and visualize the intraosseous
Effects of gelatins on calcium phosphate precipitation: a possible application for distinguishing bovine bone gelatin from porcine skin gelatin.
Hidaka S and Liu SY.
J. Food Compos. Anal., 16(4), 477-483 (2003)
Haijiang Dong et al.
Bioactive materials, 18, 164-177 (2022-04-08)
A personalized medication regimen provides precise treatment for an individual and can be guided by pre-clinical drug screening. The economical and high-efficiency simulation of the liver tumor microenvironment (TME) in a drug-screening model has high value yet challenging to accomplish.
Xingang Wang et al.
Frontiers in bioengineering and biotechnology, 9, 792893-792893 (2021-12-24)
Cardiovascular diseases have become a major threat to human health. The adhesion formation is an inevitable pathophysiological event after cardiac surgery. We have previously shown that gelatin/polycaprolactone (GT/PCL, mass ratio 50:50) electrospun nanofibrous membranes have high potential in preventing postoperative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service