Skip to Content
Merck
All Photos(3)

Key Documents

V900699

Sigma-Aldrich

Progesterone

Vetec, reagent grade, 98%

Synonym(s):

4-Pregnene-3,20-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
Inquire


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1915950
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:


Check Cart for Availability

Request a Bulk Order

grade

reagent grade

product line

Vetec

Assay

98%

mp

128-132 °C (lit.)

SMILES string

CC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17+,18-,19-,20-,21+/m0/s1

InChI key

RJKFOVLPORLFTN-LEKSSAKUSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation.
Induces maturation and secretory activity of the uterine endothelium; suppresses ovulation. Progesterone is implicated in the etiology of breast cancer.
Progesterone is a hormone that maintains pregnancy. It is essential for embryo formation, development and survival in almost all mammals. This hormone exhibits its action via progesterone receptor, which is specifically expressed in the endometrial stroma throughout pregnancy.[1] It is known to prevent preterm delivery, when administered intramuscularly.[2]

Other Notes

Steroid hormone produced by the corpus luteum.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Prophylactic administration of progesterone by vaginal suppository to reduce the incidence of spontaneous preterm birth in women at increased risk: a randomized placebo-controlled double-blind study.
da Fonseca E B, et al.
American Journal of Obstetrics and Gynecology, 188(2), 419-424 (2003)
Biology of progesterone action during pregnancy recognition and maintenance of pregnancy.
Spencer T E and Bazer F W
Frontiers in Bioscience, 7(1-3), d1879-d1898 (2002)
Nina Lisofsky et al.
NeuroImage, 118, 154-162 (2015-06-10)
Hippocampal volume has been shown to be sensitive to variations in estrogen and progesterone levels across rodents' estrous cycle. However, little is known about the covariation of hormone levels and brain structure in the course of the human menstrual cycle.
Rudolf Kaaks et al.
International journal of cancer, 134(8), 1947-1957 (2013-10-25)
Results from prospective studies on premenopausal serum hormone levels in relation to breast cancer risk have been inconclusive, especially with regard to tumor subtypes. Using a case-control study nested within the prospective European Prospective Investigation into Cancer and Nutrition (EPIC)
Cathrin Brisken
Nature reviews. Cancer, 13(6), 385-396 (2013-05-25)
Understanding the biology of the breast and how ovarian hormones impinge on it is key to rational new approaches in breast cancer prevention and therapy. Because of the success of selective oestrogen receptor modulators (SERMs), such as tamoxifen, and aromatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service