Skip to Content
Merck
All Photos(2)

Key Documents

V900425

Sigma-Aldrich

3,4-Dihydroxy-L-phenylalanine

Vetec, reagent grade, 98%

Synonym(s):

3-(3,4-Dihydroxyphenyl)-L-alanine, L-3-Hydroxytyrosine, L-DOPA, Levodopa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
197.19
Beilstein:
2215169
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

276-278 °C (lit.)

SMILES string

N[C@@H](Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1

InChI key

WTDRDQBEARUVNC-LURJTMIESA-N

Gene Information

human ... DRD3(1814)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molly J Crockett et al.
Current biology : CB, 25(14), 1852-1859 (2015-07-07)
An aversion to harming others is a core component of human morality and is disturbed in antisocial behavior. Deficient harm aversion may underlie instrumental and reactive aggression, which both feature in psychopathy. Past work has highlighted monoaminergic influences on aggression
Ezekiel J Maier et al.
Genome research, 25(5), 690-700 (2015-02-04)
Key steps in understanding a biological process include identifying genes that are involved and determining how they are regulated. We developed a novel method for identifying transcription factors (TFs) involved in a specific process and used it to map regulation
A P de Jong et al.
Clinica chimica acta; international journal of clinical chemistry, 171(1), 49-61 (1988-01-15)
L-3-(3,4-Dihydroxyphenyl)alanine (DOPA) and its 3-O-methyl metabolite (OMD) were measured in plasma and cerebrospinal fluid by a new assay which combines N,O-acetylation of amino acids in aqueous media, preparation of pentafluorobenzyl esters under anhydrous conditions, and analysis by gas chromatography-electron capture
Roshan Cools
Neuroscience and biobehavioral reviews, 30(1), 1-23 (2005-06-07)
It is well recognised that patients with Parkinson's disease exhibit cognitive deficits, even in the earliest disease stages. Whereas, L-DOPA therapy in early Parkinson's disease is accepted to improve the motor symptoms, the effects on cognitive performance are more complex:
Nicolas Morin et al.
Experimental neurology, 256, 105-116 (2013-01-31)
The treatment of motor symptoms of Parkinson disease (PD) with the dopamine (DA) precursor, l-3,4-dihydroxyphenylalanine (l-DOPA) introduced 50years ago still remains a very effective medication. However, involuntary movements termed l-DOPA-induced dyskinesias (LID) appear in the vast majority of PD patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service