Skip to Content
Merck
All Photos(1)

Key Documents

V900396

Sigma-Aldrich

Histamine

Vetec, reagent grade, ≥97%

Synonym(s):

2-(4-Imidazolyl)ethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire
5 G
Inquire


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
Inquire
5 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
Beilstein:
2012
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:


Estimated to ship on24 May 2025


Request a Bulk Order

grade

reagent grade

product line

Vetec

Assay

≥97%

bp

167 °C/0.8 mmHg (lit.)

mp

83-84 °C (lit.)

storage temp.

−20°C

SMILES string

NCCc1c[nH]cn1

InChI

1S/C5H9N3/c6-2-1-5-3-7-4-8-5/h3-4H,1-2,6H2,(H,7,8)

InChI key

NTYJJOPFIAHURM-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2 activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.
Endogenous H1 and H2 histamine receptor agonist; activates nitric oxide synthetase; potent vasodilator.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pertti Panula et al.
Nature reviews. Neuroscience, 14(7), 472-487 (2013-06-21)
Histamine acts as a modulatory neurotransmitter in the mammalian brain. It has an important role in the maintenance of wakefulness, and dysfunction in the histaminergic system has been linked to narcolepsy. Recent evidence suggests that aberrant histamine signalling in the
Lena Böhn et al.
The American journal of gastroenterology, 108(5), 634-641 (2013-05-07)
Despite the fact that food and diet are central issues, that concern patients with irritable bowel syndrome (IBS), the current understanding about the association between the intake of certain foods/food groups and the gastrointestinal (GI) symptom pattern, psychological symptoms, and
Marieke Smit et al.
The Journal of pharmacology and experimental therapeutics, 348(2), 303-310 (2013-12-07)
The novel once-daily β₂-agonist bronchodilator drug olodaterol has recently been shown to be effective in patients with allergic asthma for >24 hours. An increased cholinergic tone common to these patients may decrease the effectiveness of β₂-agonists. This could provide a
Franziska Glatzer et al.
The Journal of allergy and clinical immunology, 132(6), 1358-1367 (2013-08-13)
Epidermal hyperproliferation resulting in acanthosis is an important clinical observation in patients with atopic dermatitis, and its underlying mechanisms are not completely understood. Because increased levels of histamine are present in lesional skin, we investigated the effect of histamine, especially
Andrea Strasser et al.
Trends in pharmacological sciences, 34(1), 13-32 (2012-12-12)
Histamine is a biogenic amine that exerts its biological effects as a neurotransmitter and local mediator via four histamine receptor (HR) subtypes (H(x)Rs) - H(1)R, H(2)R, H(3)R, and H(4)R - belonging to the superfamily of G-protein-coupled receptors (GPCRs). All four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service