Skip to Content
Merck
All Photos(2)

Key Documents

V900391

Sigma-Aldrich

Thymidine

Vetec, reagent grade, 99%

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
Inquire
5 G
Inquire


Estimated to ship on24 May 2025



Select a Size

Change View
1 G
Inquire
5 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Beilstein:
89285
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:


Estimated to ship on24 May 2025


grade

reagent grade

product line

Vetec

Assay

99%

form

powder

mp

186-188 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1

InChI key

IQFYYKKMVGJFEH-XLPZGREQSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward J Gane et al.
Gastroenterology, 146(1), 138-146 (2013-09-27)
There is a close relationship between chronic hepatitis B virus infection and chronic renal disease. We analyzed changes in renal function using different markers of glomerular filtration rate (GFR) in multiple studies of telbivudine treatment of patients with chronic hepatitis
Cédric F Invernizzi et al.
The Journal of antimicrobial chemotherapy, 68(10), 2192-2196 (2013-06-12)
We recently reported the preferential selection of the K65R resistance mutation in subtype C HIV-1 compared with subtype B and showed the underlying mechanism to be dependent on subtype C-specific silent nucleotide polymorphisms, i.e. genomic mutations that change the genotype
Anne P Bouvy et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 29(8), 1587-1597 (2014-04-29)
To evaluate the influence of induction therapy on Tregs we investigated their origin, kinetics and function in kidney transplant patients after treatment with T-cell depleting rabbit antithymocyte globulin (rATG) or IL-2 receptor antagonist basiliximab. Flow cytometry was used to study
Yee Suan Poo et al.
PLoS neglected tropical diseases, 8(12), e3354-e3354 (2014-12-05)
The recent epidemic of the arthritogenic alphavirus, chikungunya virus (CHIKV) has prompted a quest to understand the correlates of protection against virus and disease in order to inform development of new interventions. Herein we highlight the propensity of CHIKV infections
Michael A Johnson et al.
Biochimica et biophysica acta, 1833(10), 2334-2347 (2013-06-19)
Actin, a constituent of the cytoskeleton, is now recognized to function in the nucleus in gene transcription, chromatin remodeling and DNA replication/repair. Actin shuttles in and out of the nucleus through the action of transport receptors importin-9 and exportin-6. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service