Skip to Content
Merck
All Photos(2)

Key Documents

V900246

Sigma-Aldrich

Maleic acid

Vetec, reagent grade, 98%

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

mp

130-135 °C (lit.)

solubility

water: 333 mg/mL, clear, colorless

density

1.59 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Chia Huang et al.
ACS applied materials & interfaces, 7(45), 25259-25269 (2015-10-27)
We have successfully introduced a proton-induced controlled reaction of HAuCl4 and poly(styrene-alt-maleic acid) (PSMA) sodium salt to prepare triangular and multicore Au@polymer nanoparticles (NPs). The interparticle interactions in the core gave rise to an absorption band at the near-infrared wavelength.
J M Herreras et al.
Ophthalmology, 99(7), 1082-1088 (1992-07-01)
This study was undertaken to see whether long-term locally applied ocular medications produced any alterations in the ocular surface, and, in particular, whether it caused damage to the mucus layer of the tear film. The authors studied the ocular surface
Hanna Kotikoski et al.
Acta ophthalmologica Scandinavica, 81(4), 355-360 (2003-07-16)
To investigate whether a nitric oxide donor given as a single oral dose is able to modify aqueous humour flow in healthy volunteers. Ten healthy volunteers participated in a randomized, double-masked and placebo-controlled cross-over study. Aqueous humour flow was assessed
K Grundke et al.
Advances in colloid and interface science, 222, 350-376 (2014-12-10)
Contact angle hysteresis phenomena on polymer surfaces have been studied by contact angle measurements using sessile liquid droplets and captive air bubbles in conjunction with a drop shape method known as Axisymmetric Drop Shape Analysis - Profile (ADSA-P). In addition
Theresia Klose et al.
Colloids and surfaces. B, Biointerfaces, 51(1), 1-9 (2006-06-27)
The adsorption of human serum albumin (HSA) and lysozyme (LSZ) on pure as well as maleic acid (MA) copolymer coated spherical soda lime glass particles was investigated under flowing conditions. Coating the glass particles with two different maleic acid copolymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service