Skip to Content
Merck
All Photos(1)

Key Documents

V900214

Sigma-Aldrich

Phosphotungstic acid hydrate

Vetec, reagent grade

Synonym(s):

Tungstophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(W3O10)4] · xH2O
CAS Number:
Molecular Weight:
2880.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

form

powder

mp

~95 °C (lit.)

solubility

water: 0.1 g/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

InChI

1S/H3O4P.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

AVFBYUADVDVJQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D van Berlo et al.
Archives of toxicology, 88(9), 1725-1737 (2014-03-26)
There is increasing concern about the toxicity of inhaled multi-walled carbon nanotubes (MWCNTs). Pulmonary macrophages represent the primary cell type involved in the clearance of inhaled particulate materials, and induction of apoptosis in these cells has been considered to contribute
Suyitno Suyitno et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 148, 99-104 (2015-04-16)
The present article reports on the enhancement of the performance and stability of natural dye-based dye-sensitized solar cells (DSSCs). Natural dyes extracted from papaya leaves (PL) were investigated as sensitizers in TiO2-based DSSCs and evaluated in comparison with N719 dye.
R Rodríguez-Gómez et al.
Talanta, 130, 561-570 (2014-08-28)
In recent decades, in parallel to industrial development, a large amount of new chemicals have emerged that are able to produce disorders in human endocrine system. These groups of substances, so-called endocrine disrupting chemicals (EDCs), include many families of compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service