Skip to Content
Merck
All Photos(1)

Key Documents

V800422

Sigma-Aldrich

Tetrahydrofuran

≥99.5%, anhydrous, contains 250 ppm BHT as stabilizer, Vetec

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
anhydrous
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Product Name

Tetrahydrofuran, anhydrous, contains 250 ppm BHT as stabilizer, ≥99.5%

grade

anhydrous

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as stabilizer

expl. lim.

1.8-11.8 %

dilution

(for analytical testing)

refractive index

n20/D 1.407 (lit.)

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan D Pensack et al.
Journal of the American Chemical Society, 137(21), 6790-6803 (2015-05-07)
We compare the singlet fission dynamics of five pentacene derivatives precipitated to form nanoparticles. Two nanoparticle types were distinguished by differences in their solid-state order and kinetics of triplet formation. Nanoparticles that comprise primarily weakly coupled chromophores lack the bulk
Luca Chiari et al.
The Journal of chemical physics, 138(7), 074301-074301 (2013-03-01)
We present total, elastic, and inelastic cross sections for positron and electron scattering from tetrahydrofuran (THF) in the energy range between 1 and 5000 eV. Total cross sections (TCS), positronium formation cross sections, the summed inelastic integral cross sections (ICS)
Tetrahydrofuran-containing macrolides: a fascinating gift from the deep sea.
Adriana Lorente et al.
Chemical reviews, 113(7), 4567-4610 (2013-03-20)
Crispin Lichtenberg et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(44), 15797-15805 (2015-09-17)
The reactivity of the all-ferrous FeN heterocubane [Fe4 (Ntrop)4 ] (1) with i) Brønsted acids, ii) σ-donors, iii) σ-donors/π-acceptors, and iv) one-electron oxidants has been investigated (trop = 5H-dibenzo[a,d]cyclo-hepten-5-yl). 1 showed self-re-assembling after reactions with i) and proved surprisingly inert in reactions with ii)
Han Yan et al.
Advanced materials (Deerfield Beach, Fla.), 27(23), 3484-3491 (2015-05-06)
Polymer solar cells are fabricated with systematic variation of the phase purity. Photovoltaic tests demonstrate that devices with ca. 10% of mixed phases outperform pure-phase devices. Photophysical studies reveal the effects of mixed phase on charge generation and recombination. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service