Skip to Content
Merck
All Photos(1)

Key Documents

V800405

Sigma-Aldrich

Tin(II) chloride

AR, ≥99%

Synonym(s):

Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99%
grade:
AR
form:
crystalline

grade

AR

vapor pressure

33 hPa (~429 °C)

product line

Vetec

Assay

≥99%

form

crystalline

pH

2.18 (20 °C)

bp

652 °C (lit.)

mp

246 °C (lit.)

anion traces

sulfate (SO42-): ≤0.01%

SMILES string

Cl[SnH2]Cl

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Hsin Hsu et al.
Nuclear medicine and biology, 41(9), 765-771 (2014-07-17)
Liposome in delivering radionuclide for cancer therapy has been expansively studied; however, liposome itself can be deliberately entrapped and destroyed by the reticuloendothelial system, causing an insufficiency of the drug delivery, which in turn would restrict the effectiveness of the
Shi Gao et al.
Clinical nuclear medicine, 39(1), e14-e19 (2013-08-07)
Tc N-MPO ([Tc N(MPO)(PNP5)]: HMPO = 2-mercaptopyridine N-oxide, and PNP5 = N-ethoxyethyl-N,N-bis[2-(bis(3-methoxypropyl)phosphino)ethyl]amine) is a new Tc radiotracer useful for myocardial perfusion imaging. This study was designed to determine its pharmacokinetics and biodistribution in healthy volunteers. Ten healthy volunteers were involved
Lars Bellner et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(1), 105-115 (2014-10-25)
Heme oxygenase (HO)-2 deficiency impairs wound healing and exacerbates inflammation following injury. We examine the impact of HO-2 deficiency on macrophage function and the contribution of macrophage HO-2 to inflammatory and repair responses to injury. Corneal epithelial debridement was performed
Filippo Galli et al.
Thyroid : official journal of the American Thyroid Association, 24(8), 1297-1308 (2014-05-08)
Differentiated thyroid carcinomas originating from thyroid follicular cells are frequent tumors of the thyroid with relatively good prognosis due to improved surgical techniques and follow-up procedures. Poorly differentiated thyroid cancers, which lose iodine uptake ability, in most cases still express
Sebastiaan Wesseling et al.
British journal of pharmacology, 172(6), 1607-1619 (2014-03-07)
Whether NO, carbon monoxide (CO) and hydrogen sulfide (H2 S) compensate for each other when one or more is depleted is unclear. Inhibiting NOS causes hypertension and kidney injury. Both global depletion of H2 S by cystathionine γ-lyase (CSE) gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service