Skip to Content
Merck
All Photos(1)

Key Documents

V800371

Sigma-Aldrich

Sodium carbonate

LR, ≥99%

Synonym(s):

Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
EC Number:
MDL number:
UNSPSC Code:
12352302
grade:
LR
form:
powder

grade

LR

product line

Vetec

Assay

≥99%

form

powder

pH

12 (25 °C, 106 g/L)

mp

851 °C (lit.)

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Waleri Root et al.
Polymers, 11(5) (2019-05-06)
Integration of sensors in textile garments requires the development of flexible conductive structures. In this work, cellulose-based woven lyocell fabrics were coated with copper during an electroless step, produced at 0.0284 M copper sulfate pentahydrate, 0.079 M potassium hydrogen L-tartrate
Eduardo Isanta et al.
Water research, 80, 149-158 (2015-05-23)
Partial nitritation for a low-strength wastewater at low temperature was stably achieved in an aerobic granular reactor. A bench-scale granular sludge bioreactor was operated in continuous mode treating an influent of 70 mg N-NH4(+) L(-1) to mimic pretreated municipal nitrogenous
Kun-Hui Lu et al.
Journal of leukocyte biology, 98(3), 301-311 (2015-05-23)
Understanding the molecular mechanisms regulating T cell reactivity is required for successful reprogramming of immune responses in medical conditions, characterized by dysfunctions of the immune system. Nck proteins are cytoplasmic adaptors mediating diverse cellular functions, including TCR signaling. By enhancing
Dries Vandamme et al.
Bioresource technology, 105, 114-119 (2011-12-21)
Microalgae hold great potential as a feedstock for biofuels or bulk protein or treatment of wastewater or flue gas. Realising these applications will require the development of a cost-efficient harvesting technology. Here, we explore the potential of flocculation induced by
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service