Skip to Content
Merck
All Photos(1)

Key Documents

V800336

Sigma-Aldrich

Potassium sodium tartrate tetrahydrate

AR, ≥99%

Synonym(s):

L(+)-Tartaric acid potassium Sodium salt, Rochelle salt, Seignette salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOCOCH(OH)CH(OH)COONa · 4H2O
CAS Number:
Molecular Weight:
282.22
Beilstein:
6113568
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay:
≥99%
grade:
AR
form:
crystalline powder
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.0005%
NH4+: ≤0.005%

SMILES string

O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.K.Na.4H2O/c5-1(3(7)8)2(6)4(9)10;;;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;4*1H2/q;2*+1;;;;/p-2/t1-,2-;;;;;;/m1....../s1

InChI key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ghina Ali et al.
Biotechnology progress, 31(4), 883-889 (2015-04-29)
The immobilization of pullulanase from Klebsiella pneumoniae by grafting was investigated. Pullulanase was linked after activation of alginate via a covalent bond between the amine groups of the enzyme and the carboxylic acid groups of alginate. The immobilization yield was
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the
Kevser Sözgen Başkan et al.
Talanta, 147, 162-168 (2015-11-26)
As the concentration of reducing sugars (RS) is controlled by European legislation for certain specific food and beverages, a simple and sensitive spectrophotometric method for the determination of RS in various food products is proposed. The method is based on
W F Tonnis et al.
Molecular pharmaceutics, 12(3), 684-694 (2015-01-13)
Protein-based biopharmaceuticals are generally produced as aqueous solutions and stored refrigerated to obtain sufficient shelf life. Alternatively, proteins may be freeze-dried in the presence of sugars to allow storage stability at ambient conditions for prolonged periods. However, to act as
Liliana Iauk et al.
Journal of enzyme inhibition and medicinal chemistry, 30(3), 360-365 (2014-07-18)
The aim of this study was to examine the bioactivity of the methanol fraction (MF) and n-hexane fraction (HF) of Thymus capitatus leaves in relation to their constituents analysed by gas chromatography and gas chromatography-mass spectrometry. The effects of T.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service