Skip to Content
Merck
All Photos(2)

Key Documents

V800334

Sigma-Aldrich

Potassium persulfate

LR, ≥98%

Synonym(s):

Potassium peroxodisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O8
CAS Number:
Molecular Weight:
270.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder
Pricing and availability is not currently available.

grade

LR

vapor density

9.3 (vs air)

product line

Vetec

Assay

≥98%

form

crystalline powder

pH

2.5-4.5 (25 °C, 27 g/L)

anion traces

chloride (Cl-): ≤0.04%

cation traces

Fe: ≤0.005%

SMILES string

[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

USHAGKDGDHPEEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huan Niu et al.
Biosensors & bioelectronics, 26(7), 3175-3180 (2011-01-18)
A novel label free electrochemiluminescence (ECL) immunosensor based on the ECL of peroxydisulfate solution for detection of α-1-fetoprotein (AFP) has been developed. For this proposed immunosensor, L-cysteine was firstly electrodeposited on the gold electrode surface, which promoted the electron transfer
Bernaurdshaw Neppolian et al.
Water research, 44(12), 3687-3695 (2010-04-27)
The sonochemical oxidation of As(III) in the presence of peroxydisulfate ion (PDS) has been investigated. Sulfate anion radicals and OH radicals produced during acoustic cavitation readily oxidized As(III) to As(V) in an aqueous environment. The rate of oxidation of As(III)
Harriet Mörtstedt et al.
Rapid communications in mass spectrometry : RCM, 25(2), 327-340 (2010-12-31)
Oxidative stress has been suggested as an underlying mechanism of many human diseases. However, definitive evidence for this association has not been presented due to different shortcomings of the methods used to measure biomarkers of oxidative stress. Persulfates are oxidizing
Zhiyong Yang et al.
The Journal of organic chemistry, 77(16), 7086-7091 (2012-07-28)
Nontransition metal-catalyzed synthesis of 2-aryl benzothiazoles was achieved through K(2)S(2)O(8)-mediated oxidative condensation of benzothiazoles with aryl aldehydes. The same transformation can also be effected when the aryl aldehydes were replaced with phenylglyoxylic acids.
Xiuping Zhu et al.
Environmental science & technology, 48(12), 7157-7163 (2014-05-28)
Several technologies, including pressure-retarded osmosis (PRO), reverse electrodialysis (RED), and capacitive mixing (CapMix), are being developed to recover energy from salinity gradients. Here, we present a new approach to capture salinity gradient energy based on the expansion and contraction properties

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service