Skip to Content
Merck
All Photos(1)

Key Documents

V800320

Sigma-Aldrich

Potassium hydroxide

AR, flakes, ≥85%

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
grade:
AR
form:
flakes
solubility:
H2O: soluble 1120 g/L
Pricing and availability is not currently available.

grade

AR

vapor pressure

1 mmHg ( 719 °C)

product line

Vetec

Assay

≥85%

form

flakes

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

solubility

H2O: soluble 1120 g/L

anion traces

carbonate (CO32-): ≤1.00%
chloride (Cl-): ≤0.0020%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.0030%

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mizuki Takemoto et al.
PloS one, 10(6), e0131094-e0131094 (2015-06-27)
Channelrhodopsin (ChR) is a light-gated cation channel that responds to blue light. Since ChR can be readily expressed in specific neurons to precisely control their activities by light, it has become a powerful tool in neuroscience. Although the recently solved
Ashok R Patel et al.
Journal of agricultural and food chemistry, 63(19), 4862-4869 (2015-05-02)
The aim of this study was to use a detailed rheological characterization to gain new insights into the gelation behavior of natural waxes. To make a comprehensive case, six natural waxes (differing in the relative proportion of chemical components: hydrocarbons
K Y Foo et al.
Bioresource technology, 130, 696-702 (2013-01-22)
In this work, preparation of granular activated carbon from oil palm biodiesel solid residue, oil palm shell (PSAC) by microwave assisted KOH activation has been attempted. The physical and chemical properties of PSAC were characterized using scanning electron microscopy, volumetric
Peng Li et al.
Angewandte Chemie (International ed. in English), 54(2), 574-577 (2014-11-15)
Self-assembly of a trigonal building subunit with diaminotriazines (DAT) functional groups leads to a unique rod-packing 3D microporous hydrogen-bonded organic framework (HOF-3). This material shows permanent porosity and demonstrates highly selective separation of C2H2/CO2 at ambient temperature and pressure.
Nilesh M Agalave et al.
Pain, 155(9), 1802-1813 (2014-06-24)
Extracellular high mobility group box-1 protein (HMGB1) plays important roles in the pathogenesis of nerve injury- and cancer-induced pain. However, the involvement of spinal HMGB1 in arthritis-induced pain has not been examined previously and is the focus of this study.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service