Skip to Content
Merck
All Photos(1)

Key Documents

V800280

Sigma-Aldrich

Ninhydrin

AR, ≥99%

Synonym(s):

1,2,3-Indantrione monohydrate, 2,2-Dihydroxy-1,3-indanedione, Trioxohydrindene monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Beilstein:
1910963
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

AR

product line

Vetec

Assay

≥99%

mp

250 °C (dec.) (lit.)

functional group

hydroxyl
ketone

SMILES string

OC1(O)C(=O)c2ccccc2C1=O

InChI

1S/C9H6O4/c10-7-5-3-1-2-4-6(5)8(11)9(7,12)13/h1-4,12-13H

InChI key

FEMOMIGRRWSMCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for the detection of free amino groups in amino acids, peptides and proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Feng Xie et al.
Journal of neurochemistry, 122(3), 605-618 (2012-05-23)
Neurons located in the trigeminal subnucleus caudalis (Vc) play crucial roles in pain and sensorimotor functions in the orofacial region. Because of many anatomical and functional similarities with the spinal dorsal horn (SDH), Vc has been termed the medullary dorsal
Joseph Almog et al.
Journal of forensic sciences, 56 Suppl 1, S162-S165 (2011-01-14)
We explored the quality distribution of ninhydrin-developed prints on A4 bond paper in two groups of individuals, in Israel and in India. While the quality distributions of the developed marks in both countries had some dissimilarities, both groups showed the
Mendel Friedman
Journal of agricultural and food chemistry, 52(3), 385-406 (2004-02-05)
The reaction of ninhydrin with primary amino groups to form the purple dye now called Ruhemann's purple (RP) was discovered by Siegfried Ruhemann in 1910. In addition, imines such as pipecolic acid and proline, the guanidino group of arginine, the
Chao Li et al.
Journal of biomaterials science. Polymer edition, 23(1-4), 405-424 (2011-02-12)
Biomaterials have been playing important roles in cartilage regeneration. Although many scaffolds have been reported to enhance cartilage regeneration, none of the scaffolds available are optimal regarding mechanical properties, integration with host cartilage and providing proper micro-environment for chondrocyte attachment
Jianhui Gu et al.
Journal of tissue engineering and regenerative medicine, 6(2), 163-168 (2011-03-04)
This paper describes a clinical case study in which a chitosan/polyglycolic acid nerve guidance conduit (chitosan-PGA NGC) was utilized to repair a 30 mm long median nerve defect in the right distal forearm of a 55 year-old male patient. Thirty-six

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service