Skip to Content
Merck
All Photos(2)

Key Documents

V800217

Sigma-Aldrich

Iodine

AR, ≥99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:
grade:
AR
form:
solid
Pricing and availability is not currently available.

grade

AR

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

product line

Vetec

Assay

≥99.8%

form

solid

resistivity

1.3E15 μΩ-cm

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eike I Piechowiak et al.
Radiology, 275(3), 692-697 (2015-02-06)
To determine the effect of the use of iodinated contrast agents on the formation of DNA double-strand breaks during chest computed tomography (CT). This study was approved by the institutional review board, and written informed consent was obtained from all
Michael B Zimmermann
Thyroid : official journal of the American Thyroid Association, 17(9), 829-835 (2007-10-25)
Iodine is required for the production of thyroid hormones, which are essential for normal brain development, and the fetus, newborn, and young child are particularly vulnerable to iodine deficiency. The iodine requirement increases during pregnancy and recommended intakes are in
Michael B Zimmermann et al.
Nutrition reviews, 70(10), 553-570 (2012-10-06)
Iodine status has been historically assessed by palpation of the thyroid and reported as goiter rates. Goiter is a functional biomarker that can be applied to both individuals and populations, but it is subjective. Iodine status is now assessed using
Tim I M Korevaar et al.
The Journal of clinical endocrinology and metabolism, 98(11), 4382-4390 (2013-09-17)
Premature delivery is an important risk factor for child mortality and psychiatric, metabolic, and cardiovascular disease later in life. In the majority of cases, the cause of prematurity cannot be identified. Currently, it remains controversial whether abnormal maternal thyroid function
Zulfiqar A Bhutta et al.
Lancet (London, England), 382(9890), 452-477 (2013-06-12)
Maternal undernutrition contributes to 800,000 neonatal deaths annually through small for gestational age births; stunting, wasting, and micronutrient deficiencies are estimated to underlie nearly 3·1 million child deaths annually. Progress has been made with many interventions implemented at scale and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service