Skip to Content
Merck
All Photos(1)

Documents

V800190

Sigma-Aldrich

Formaldehyde solution

LR, contains 5-8% methanol as stabilizer, 37-41 % (w/v)

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

grade

LR

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

product line

Vetec

Assay

37.0-41.0% (acidimetric)

form

liquid

autoignition temp.

572 °F

contains

5-8% methanol as stabilizer

concentration

37-41 % (w/v)

density

1.09 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use (Laboratory chemicals)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

147.2 °F

Flash Point(C)

64 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie Belloc et al.
Fertility and sterility, 101(6), 1588-1593 (2014-04-03)
To evaluate sperm DNA fragmentation in normozoospermic male partners of couples undergoing infertility evaluation. Retrospective cohort study. Clinical andrology laboratory. A total of 1,974 consecutive normozoospermic men selected from a larger cohort of 4,345 consecutive, nonazoospermic men presenting for infertility
Arpeeta Sharma et al.
Diabetes, 64(11), 3937-3950 (2015-06-28)
Patients with diabetes have an increased risk of developing atherosclerosis. Endothelial dysfunction, characterized by the lowered bioavailability of endothelial NO synthase (eNOS)-derived NO, is a critical inducer of atherosclerosis. However, the protective aspect of eNOS in diabetes-associated atherosclerosis remains controversial
Silvia Surinova et al.
EMBO molecular medicine, 7(9), 1166-1178 (2015-08-09)
Non-invasive detection of colorectal cancer with blood-based markers is a critical clinical need. Here we describe a phased mass spectrometry-based approach for the discovery, screening, and validation of circulating protein biomarkers with diagnostic value. Initially, we profiled human primary tumor
James Larkin et al.
The New England journal of medicine, 373(1), 23-34 (2015-06-02)
Nivolumab (a programmed death 1 [PD-1] checkpoint inhibitor) and ipilimumab (a cytotoxic T-lymphocyte-associated antigen 4 [CTLA-4] checkpoint inhibitor) have been shown to have complementary activity in metastatic melanoma. In this randomized, double-blind, phase 3 study, nivolumab alone or nivolumab plus
G R Moore et al.
Neurology, 55(10), 1506-1510 (2000-11-30)
To determine the pathologic basis of areas not exhibiting signal of the short-T2 component of the T2 relaxation distribution in MS, as studied in formalin-fixed brain. A myelin-specific MRI signal would be of great importance in assessing demyelination in patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service