Skip to Content
Merck
All Photos(1)

Key Documents

V800180

Sigma-Aldrich

Ethylene glycol

AR, ≥99%

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
AR
bp:
195-198 °C
vapor pressure:
0.08 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

AR

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

752 °F

expl. lim.

15.3 %

dilution

(for analytical testing)

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Wang et al.
Journal of the American Chemical Society, 135(5), 1941-1951 (2013-01-16)
This article describes a robust method for the facile synthesis of small Ag nanocubes with edge lengths controlled in the range of 18-32 nm. The success of this new method relies on the substitution of ethylene glycol (EG)--the solvent most
Revital Goldshmid et al.
Methods (San Diego, Calif.), 84, 35-43 (2015-05-02)
A method for the preparation of suspension culture microcapsules used in the bioprocessing of human mesenchymal stem cells (hMSCs) is reported. The microcapsules are prepared from a semi-synthetic hydrogel comprising Pluronic®F127 conjugated to denatured fibrinogen. The Pluronic-fibrinogen adducts display a
Laetitia Galea et al.
Biomaterials, 34(27), 6388-6401 (2013-06-13)
Calcium phosphates (CaPs) are widely used as bone graft substitutes but are inherently brittle, hence restricting their use to mechanically protected environments. Combining them with a tough polymer matrix could potentially lead to a composite with load-bearing properties. However, the
Birgit Oberleitner et al.
Chemical communications (Cambridge, England), 49(16), 1615-1617 (2013-01-23)
This work describes an integrated approach for designing on demand Self-Assembled Monolayers (SAMs) on silicon oxides and particularly glass substrates for cell biology applications. Starting from commercially available compounds, the strategy relies on thiol-ene reaction and provides high quality SAMs
Peter Mygind Leth et al.
Forensic science international, 155(2-3), 179-184 (2005-10-18)
Ethylene glycol (EG) can be found in many agents, such as antifreeze. Ingestion of EG may cause serious poisoning. Adults are typically exposed when EG is ingested as a cheap substitute for ethanol or in suicide-attempts. Children may be exposed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service