Skip to Content
Merck
All Photos(1)

Key Documents

V800141

Sigma-Aldrich

D-(−)-Fructose

LR

Synonym(s):

D-Levulose, Fruit sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1239004
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

grade

LR

product line

Vetec

Assay

≥98%

mp

119-122 °C (dec.) (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1

InChI key

BJHIKXHVCXFQLS-UYFOZJQFSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B P Chumpitazi et al.
Alimentary pharmacology & therapeutics, 42(4), 418-427 (2015-06-25)
A low fermentable oligosaccharides, disaccharides, monosaccharides and polyols (FODMAP) diet can ameliorate symptoms in adult irritable bowel syndrome (IBS) within 48 h. To determine the efficacy of a low FODMAP diet in childhood IBS and whether gut microbial composition and/or metabolic
Souhir Marsit et al.
Molecular biology and evolution, 32(7), 1695-1707 (2015-03-10)
Although an increasing number of horizontal gene transfers have been reported in eukaryotes, experimental evidence for their adaptive value is lacking. Here, we report the recent transfer of a 158-kb genomic region between Torulaspora microellipsoides and Saccharomyces cerevisiae wine yeasts
Marine Zwicke et al.
Annals of botany, 116(6), 1001-1015 (2015-04-09)
Extreme climatic events such as severe droughts are expected to increase with climate change and to limit grassland perennity. The present study aimed to characterize the adaptive responses by which temperate herbaceous grassland species resist, survive and recover from a
K Shiraga et al.
Carbohydrate research, 406, 46-54 (2015-02-07)
Terahertz time-domain attenuated total reflection measurements of monosaccharide (glucose and fructose) and disaccharide (sucrose and trehalose) solutions from 0.146 M to 1.462 M were performed to evaluate (1) the hydration state and (2) the destructuring effect of saccharide solutes on
Carola H Blessing et al.
Tree physiology, 35(6), 585-598 (2015-04-17)
The response of carbon allocation to drought has often been studied in terms of short-term transport velocity of recently fixed carbon from leaves to roots and root respiration. However, its dynamic response to other environmental conditions, e.g., to changes in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service