Skip to Content
Merck
All Photos(1)

Key Documents

V800139

Sigma-Aldrich

Cyclohexanone

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
AR
bp:
155 °C (lit.)
vapor pressure:
3.4 mmHg ( 20 °C)

grade

AR

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

dilution

(for analytical testing)

refractive index

n20/D 1.450 (lit.)

pH

~7 (20 °C, 70 g/L)

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of fragment screening and merging to the discovery of inhibitors of the Mycobacterium tuberculosis cytochrome P450 CYP121.
Sean A Hudson et al.
Angewandte Chemie (International ed. in English), 51(37), 9311-9316 (2012-08-15)
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2629-2637 (2012-09-07)
Iron (and to a lesser extent manganese) in the wall of a 316 stainless steel (SS) reactor is responsible for the hydrogenation of cyclohexanone to cyclohexanol when using an aqueous formic acid solution under high temperature and pressure water (HTPW)
Michael Eddleston et al.
Toxicology, 294(2-3), 94-103 (2012-03-01)
Organophosphorus (OP) insecticide self-poisoning is responsible for about one-quarter of global suicides. Treatment focuses on the fact that OP compounds inhibit acetylcholinesterase (AChE); however, AChE-reactivating drugs do not benefit poisoned humans. We therefore studied the role of solvent coformulants in
Annhelen Lu et al.
Chemical communications (Cambridge, England), 48(78), 9699-9701 (2012-08-23)
The use of functional core-shell micelles as asymmetric catalytic nanoreactors for organic reactions in water is presented. An unprecedented increase in rate of reaction was achieved, which is proposed to be associated with the ability of the nanostructures to effectively
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2638-2643 (2012-09-07)
The processing of renewable feedstocks to platform chemicals and, to a lesser degree, fuels is a key part of sustainable development. In particular, the combination of lignocellulosic biomass with hydrothermal upgrading (HTU), using high temperature and pressure water (HTPW), is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service