Skip to Content
Merck
All Photos(1)

Key Documents

V800114

Sigma-Aldrich

Chlorobenzene

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
AR
bp:
132 °C (lit.)
vapor pressure:
11.8 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

AR

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

1178 °F

expl. lim.

7.1 %

dilution

(for analytical testing)

refractive index

n20/D 1.524 (lit.)

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Chao Chen et al.
ACS applied materials & interfaces, 7(8), 4616-4622 (2015-02-11)
Controlled dispersion of single-walled carbon nanotubes (SWCNTs) in common solvents is a challenging issue, especially for the rising need of low cost flexible transparent conducting films (TCFs). Utilizing conductive polymer as surfactant to facilitate SWCNTs solubility is the most successful
Kyeongil Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(7), 1241-1247 (2015-01-13)
Fully printed perovskite solar cells are demonstrated with slot-die coating, a scalable printing method. A sequential slot-die coating process is developed to produce efficient perovskite solar cells and to be used in a large-scale roll-to-roll printing process. All layers excluding
Kwanpyo Kim et al.
Nature communications, 5, 4781-4781 (2014-09-03)
One-dimensional defects in graphene have a strong influence on its physical properties, such as electrical charge transport and mechanical strength. With enhanced chemical reactivity, such defects may also allow us to selectively functionalize the material and systematically tune the properties
Chuan-Yu Qin et al.
Water environment research : a research publication of the Water Environment Federation, 85(2), 133-140 (2013-03-12)
A two-dimensional laboratory sand tank was installed to study the remediation efficiency of surfactant-enhanced air sparging (-SEAS) coupled with soil vapor extraction (SVE) in nonaqueous phase liquid (NAPL) polluted sites. During initial stages of remediation, it was more reasonable to
Kerry N McPhedran et al.
Water research, 47(7), 2222-2230 (2013-03-12)
The sorption behaviour of hydrophobic organic compounds (HOCs) 1,2,4,5-tetrachlor-obenzene (TeCB), pentachlorobenzene (PeCB) and hexachlorobenzene (HCB) to Aldrich humic acid (AHA) and municipal wastewater treatment plant (MWTP) influent colloidal organic carbon (COC) was investigated using the gas-stripping technique. Gas stripping assumptions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service