Skip to Content
Merck
All Photos(1)

Key Documents

V800100

Sigma-Aldrich

Calcium carbonate

LR, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

Assay

≥98.5%

form

powder

pH

8

density

2.93 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.025%
sulfate (SO42-): ≤0.3%

cation traces

As: ≤0.0003%
heavy metals (as Pb): ≤0.005%

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie A Jamal et al.
Lancet (London, England), 382(9900), 1268-1277 (2013-07-23)
Phosphate binders (calcium-based and calcium-free) are recommended to lower serum phosphate and prevent hyperphosphataemia in patients with chronic kidney disease, but their effects on mortality and cardiovascular outcomes are unknown. We aimed to update our meta-analysis on the effect of
Jane A Cauley et al.
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were
M Azizur Rahman et al.
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Ling Li et al.
Nature materials, 13(5), 501-507 (2014-04-01)
Hierarchical composite materials design in biological exoskeletons achieves penetration resistance through a variety of energy-dissipating mechanisms while simultaneously balancing the need for damage localization to avoid compromising the mechanical integrity of the entire structure and to maintain multi-hit capability. Here
Polysaccharides and proteoglycans in calcium carbonate-based biomineralization.
José L Arias et al.
Chemical reviews, 108(11), 4475-4482 (2008-07-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service