Skip to Content
Merck
All Photos(1)

Key Documents

V800065

Sigma-Aldrich

Barium chloride dihydrate

LR, 99-102%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
powder
Pricing and availability is not currently available.

grade

LR

product line

Vetec

Assay

99-102%

form

powder

pH

5-8 (25 °C, 50 g/L)

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Merriam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4614-4622 (2013-03-08)
After G-protein-coupled receptor activation and signaling at the plasma membrane, the receptor complex is often rapidly internalized via endocytic vesicles for trafficking into various intracellular compartments and pathways. The formation of signaling endosomes is recognized as a mechanism that produces
Fan Hu et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(12), 1051-1058 (2012-08-15)
Danshen (Salviae Miltiorrhizae Radix) and Gegen (Puerariae Lobatae Radix) have been widely used in treating cardiovascular diseases for thousands of years in China. The present study was carried out to evaluate the effects of a Danshen and Gegen decoction (DG)
Virginia Bay et al.
Glia, 60(4), 651-660 (2012-02-01)
Uptake of K(+) released by axons during action potential propagation is a major function of astrocytes. Here, we demonstrate the importance of glial inward rectifying potassium channels (Kir) in regulating extracellular K(+) ([K(+)](o)) and axonal electrical activity in CNS white
Hanna Heikkinen et al.
Investigative ophthalmology & visual science, 53(9), 5653-5664 (2012-06-30)
To examine the amplification and kinetics of murine rod photoresponses by recording ERG flash responses in vivo and ex vivo from the same retina. We also aimed to evaluate the two available methods for isolating the rod signal from the
Nathalie M Goodfellow et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(17), 5804-5809 (2012-04-28)
The 5-HT(5A) receptor is the least understood serotonin (5-HT) receptor. Here, we electrophysiologically identify and characterize a native 5-HT(5A) receptor current in acute ex vivo brain slices of adult rodent prefrontal cortex. In the presence of antagonists for the previously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service