Skip to Content
Merck
All Photos(1)

Documents

1193006

USP

Diethylcarbamazine citrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Diethylcarbamazine citrate salt, N,N-Diethyl-4-methyl-1-piperazinecarboxamide citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H21N3O · C6H8O7
CAS Number:
Molecular Weight:
391.42
Beilstein:
3836352
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diethylcarbamazine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1

InChI

1S/C10H21N3O.C6H8O7/c1-4-12(5-2)10(14)13-8-6-11(3)7-9-13;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-9H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

PGNKBEARDDELNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethylcarbamazine citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Diethylcarbamazine Citrate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M García-Hernández et al.
International immunopharmacology, 23(1), 113-120 (2014-08-26)
We tested whether diethylcarbamazine (DEC) or ivermectin (IVM), both antiparasitic drugs with reported immunomodulatory properties, were able to affect the immune system to potentiate host defense mechanisms and protect against actinomycetoma in a mouse model. Male BALB/c mice of 10-12
Mark S Schmidt et al.
Journal of pharmaceutical and biomedical analysis, 98, 307-310 (2014-07-01)
A sensitive and selective liquid chromatographic method using mass spectrometric detection was developed for the determination of diethylcarbamazine (DEC) in human plasma. DEC and its stable isotope internal standard d3-DEC were extracted from 0.25mL of human plasma using solid phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service