G005597
HPLC Analysis of α-Casein Tryptic Digest on BIOshell A160 Peptide C18
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
sample/matrix: tryptic digested α-casein
column: BIOshell A160 Peptide C18, 5 cm x 4.6 mm I.D., 2.7 μm particles (66913-U)
mobile phase: [A] 0.1% TFA in water; [B] 0.1% TFA in water:acetonitilre (10:90, A:B)
gradient: 0-0.05 min, 0-20% B; 0.05-0.40 min, to 40% B; 0.40-0.50 min, to 50% B; 0.50-0.69 min, to 90% B; 0.69-0.70 min, to 0% B; held for 0.3 min
flow rate: 4 mL/min
column temp.: 35 °C
detector: UV, 215 nm
injection: 2 μL
suitability
application for HPLC
application(s)
life science and biopharma
Analysis Note
Other Notes
Legal Information
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service