G005066
GC Analysis of cis/trans Limonene Oxide Enantiomers on Astec® CHIRALDEX™ B-PH
suitable for GC
About This Item
Recommended Products
technique(s)
gas chromatography (GC): suitable
test parameters
column: Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 μm (71023AST)
oven: 90 °C
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 24 psi
injection: 1 μL, 80:1 split
sample: 3 mg/mL in methylene chloride
peak 1: d-cis or d-trans limonene oxide
peak 2: l-cis and trans limonene oxide
peak 3: d-trans or d-cis limonene oxide
application(s)
chemicals and industrial polymers
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Legal Information
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service