Skip to Content
Merck
All Photos(1)

Documents

818404

TSKgel® G1000HXL (5 µm) HPLC Columns

L x I.D. 4 cm × 6 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® Multipore HXL-M guard column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® G1000HXL HPLC Column for Light Scattering Applications, phase polystyrene-divinylbenzene, L × I.D. 4 cm × 6 mm, 5 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh 18404

parameter

1.2 mL/min flow rate
60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

polymeric particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

5 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Guard Column for TSKgel MultiporeHxl-M column, 5μm

Physical form

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service