Skip to Content
Merck
All Photos(1)

Key Documents

59473

Plunger

for use with LC-10AS, Shimadzu 228-17019-93

Synonym(s):

Optimize Technologies HPLC Pump Part for Shimadzu

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

packaging

pkg of 1

manufacturer/tradename

Shimadzu 228-17019-93

compatibility

for use with LC-10AS

Looking for similar products? Visit Product Comparison Guide

Legal Information

Shimadzu is a trademark of Shimadzu Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service