Skip to Content
Merck
All Photos(16)

Documents

ZRB2292

Sigma-Aldrich

Anti-Mucin-14/EMCN Antibody, clone 1E8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Endomucin, Endomucin-2, Gastric cancer antigen Ga34, MUC-14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E8, recombinant monoclonal

description

1E8 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 27.45 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... EMCN(51705)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E8 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Mucin-14. It targets an epitope within 18 amino acids from the cytoplasmic domain from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the cytoplasmic domain within the C-terminal half of human Mucin-14.

Application

Quality Control Testing

Evaluated by Western Blotting in bEND.3 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Mucin-14/EMCN in bEND.3 cell lysate.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Mucin-14/EMCN in bEND.3 and HepG2 cells.

Affinity Binding Assay: A representative lot of this antibody bound Mucin-14/EMCN with a KD of 2.7 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Mucin-14/EMCN in human kidney tissue sections.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected Mucin-14/EMCN in one million bEND.3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Endomucin (UniProt: Q9ULC0; also known as Endomucin-2, Gastric cancer antigen Ga34, Mucin-14, MUC-14) is encoded by the EMCN (also known as EMCN2, MUC14) gene (Gene ID: 51705) in human.  Mucins are large, heavily glycosylated, extracellular proteins that establish a selective molecular barrier at the epithelial surface and engage in morphogenetic signal transduction. They are the major macromolecular components of mucus. They have a unique and characteristic sequence of tandemly repeating amino acids rich in serine and/or threonine. They are expressed by various epithelial cell types that exist in relatively harsh environments, such as air–water interface of the respiratory system and the acidic environment of the stomach. Mucin-14 is a single-pass type I membrane sialoglycoprotein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 19-190) that is highly O-glycosylated at serine and threonine residues; a transmembrane domain (aa 191-211); and a cytoplasmic domain (aa 212-261). Three isoforms of Mucin-14 have been described that are produced by alternative splicing. Isoform 1 is membrane associated protein that consistently localizes to the plasma membrane and less abundantly to Golgi-like perinuclear stacks. Isoform 2 lacks amino acids 140-222, and isoform 3 is a secretory form. Its expression is observed in heart, kidney, and lungs. It is also expressed in retinal endothelial cells, and its expression is reduced under conditions of high blood glucose in vitro and in vivo. Its overexpression can restore the glycocalyx of retinal endothelial cells in diabetic rats. In the eye, it inhibits retinal inflammation and promotes retinal microvascular angiogenesis during retinal development. It has been identified as a novel VEGF‐induced angiogenic regulator that can reduce VEGF‐induced migration, proliferation, and tube formation of human retinal microvascular endothelial cells. It is also reported to interfere with the assembly of focal adhesion complexes and inhibit the interaction between cells and the extracellular matrix. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhang, G.,  et al (2000). Animal Model Exp. Med. 3(4); 325-329; Park-Windhol, C., et al. (2017). Sci.  Rep. 7(1); 17138; Hollingsworth, MA., and Swanson, BJ. (2004). Nat. Rev. Cancer. 4(1); 45-60).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service