Skip to Content
Merck
All Photos(1)

Documents

SRP8032

Sigma-Aldrich

IL-1 Receptor Type I (human): FC (human)

recombinant, expressed in HEK 293 cells, >95% (SDS-PAGE)

Synonym(s):

Interleukin-1 receptor alpha, Interleukin-1 receptor type I, RI, Serpin F1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~80 kDa by SDS-PAGE

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL1R1(3554)

General description

IL-1RI is the receptor for interleukin-1 alpha (IL-1A), beta (IL-1B), and interleukin-1 receptor antagonist protein (IL-1RA). Binding to the agonist leads to the activation of NF-kappaB. Signaling involves formation of a ternary complex containing IL1RAP, leading the activation of TOLLIP, MYD88, and IRAK1 or IRAK2.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 50 μL deionized water.

Other Notes

Human IL-1RI (interleukin-1 receptor type I) (aa 21-336) is fused at the C-terminus to the Fc portion of human IgG1.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service