Skip to Content
Merck
All Photos(1)

Key Documents

SML3898

Sigma-Aldrich

P144 diammonium

≥95% (HPLC)

Synonym(s):

Disitertide diammonium, P144 diammonium, Peptide 144 diammonium, TSLDASIIWAMMQN diammonium, Thr-Ser-Leu-Asp-Ala-Ser-Ile-Ile-Trp-Ala-Met-Met-Gln-Asn diammonium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H109N17O22S2
Molecular Weight:
1580.82
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Human type 3 TGF-β receptor sequence-derived TGF-beta1 (TGF-β1) inhibitor that blocks TGF-β1-mediated responses in vitro and in vivo.


P144 (Disitertide) is a 14-aa peptide TGF-beta1 (TGF-β1) inhibitor whose sequence corresponds to aa731-744 of human type 3 TGF-β receptor (UniProt Q03167). P144 blocks TGF-β1 from binding the type 3 TGF-β receptor and inhibits TGF-β1-mediated responses in vitro (80% MV1Lu growth inhibition by 200 μg/mL P144 in the presence of 200 pg/mL human TGF-β1) and in vivo (57% inhibition of hepatic stellate cells (HSCs) activation in a rat model of CCl4 inhalation-induced cirrhosis/hepatic fibrosis; 70 μg P144/rat via q.o.d. i.p.).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peptide inhibitors of transforming growth factor-beta enhance the efficacy of antitumor immunotherapy
International Journal of Cancer. Journal International Du Cancer, 125(11), 2614-2623 (2009)
A synthetic peptide from transforming growth factor beta type III receptor inhibits liver fibrogenesis in rats with carbon tetrachloride liver injury
Cytokine, 22(1-2), 12-20 (2003)
Topical application of a peptide inhibitor of transforming growth factor-beta1 ameliorates bleomycin-induced skin fibrosis
The Journal of Investigative Dermatology, 125(3), 450-455 (2005)
Identification of peptide inhibitors of transforming growth factor beta 1 using a phage-displayed peptide library
Cytokine, 39(2), 106-115 (2007)
P144 a Transforming Growth Factor Beta Inhibitor Peptide, Generates Antifibrogenic Effects in a Radiotherapy Induced Fibrosis Model
Current Oncology (Toronto, Ont.), 29(4), 2650-2661 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service