Skip to Content
Merck
All Photos(1)

Key Documents

SML3002

Sigma-Aldrich

Sulfuretin

≥98% (HPLC)

Synonym(s):

Sulfuretin, Sulphuretin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
RM 809.00

RM 809.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
RM 809.00

About This Item

Empirical Formula (Hill Notation):
C15H10O5
Molecular Weight:
270.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

RM 809.00


Estimated to ship on02 June 2025


Request a Bulk Order

description

≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

OC1=CC=C2C(O/C(C2=O)=C\C3=CC(O)=C(O)C=C3)=C1

InChI

1S/C15H10O5/c16-9-2-3-10-13(7-9)20-14(15(10)19)6-8-1-4-11(17)12(18)5-8/h1-7,16-18H/b14-6-

InChI key

RGNXWPVNPFAADO-NSIKDUERSA-N

General description

Sulfuretin is an aurones flavonoid found in the extract of Rhus verniciflua. The Extract of Rhus verniciflua is known in the traditional medicine and scientific community to have anti-cancer properties.Sulfuretin, one of the major constituents of Rhus verniciflua extract was found to have anti-cancer activities via activation of caspase-3 and inhibition of PARP. Moreover, sulfuretin downregulates cyclin D proteins by increasing the expression of miR-30C, a tumor suppressor micro RNA that is studied in regulation of cell proliferation.2 Structure-activity relationships (SAR) of Sulfuretin analogs inhibits the proliferation of leukemia cancer line panel in vitro and in vivo in zebrafish and mice models.Furthermore, Sulfuretin 100mg/Kg in vivo study was beneficial in treating lymphoedema in mice.In addition, Sulfuretin has several other activities such as:
  • Sulfuretin is an anti-adipogenic agent, suppressing the adipogenic factors C/EBPα, C/EBPβ and PPARγ, which are crucial regulators at the initial step of adipogenesis.
  • Sulfuretin is a potent antioxidant which protects SH-SY5Y cell line from 6-OHDA induced ROS.
  • Sulfuretin has antiviral activity through the inhibition of influenza virus neuraminidase in IC50 of 36μM.

Biochem/physiol Actions

Anti-adipogenic, antineoplastic activity, antioxidative stress, Anti-Inflammatory

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this product safe for human consumption? If so, how many milligrams per day should be taken?

    1 answer
    1. This product is intended for research use only. It is not suitable for any other purpose, which includes, but is not limited to, unauthorized commercial use, in vitro diagnostic use, or therapeutic use in humans or animals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service