Skip to Content
Merck
All Photos(1)

Documents

SML1157

Sigma-Aldrich

Eact

≥98% (HPLC)

Synonym(s):

3,4,5-Trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-2-thiazolyl)-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O5S
CAS Number:
Molecular Weight:
428.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC(OC)=C(OC)C(OC)=C1)N(CCOC)C2=NC(C3=CC=CC=C3)=CS2

InChI

1S/C22H24N2O5S/c1-26-11-10-24(22-23-17(14-30-22)15-8-6-5-7-9-15)21(25)16-12-18(27-2)20(29-4)19(13-16)28-3/h5-9,12-14H,10-11H2,1-4H3

InChI key

ZUXNHFFVQWADJL-UHFFFAOYSA-N

Biochem/physiol Actions

Eact activates the Calcium-activated Chloride channel (CaCC) TMEM16A CaCC. Eact stimulates CaCC conductance in salivary gland cells and submucosal airway epithelial cells. The compound Eact also stimulates submucosal gland secretion in human bronchial tissue and smooth muscle contraction in mouse intestine.
In rat arterial myocytes, Eact [3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-2-thiazolyl)benzamide] helps to activate chloride current and prevents estradiol synthesis in cultured mouse granulosa cells. It is a small molecule activator of TMEM16A (transmembrane member 16A), that activates TRPV1 (transient receptor potential cation channel subfamily V member 1) and generates pain and itch related symptoms. Eact also initiates endogenous TRPV1 expressed by DRG (dorsal root ganglion) neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eact, a small molecule activator of TMEM16A, activates TRPV1 and elicits pain?and itch?related behaviours.
Liu S, et al.
British Journal of Pharmacology, 173(7), 1208-1218 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service