Skip to Content
Merck
All Photos(6)

Key Documents

SAB4503787

Sigma-Aldrich

Anti-phospho-Tyrosine Hydroxylase (pSer19) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
RM 2,713.00

RM 2,713.00


Estimated to ship on01 June 2025



Select a Size

Change View
100 μG
RM 2,713.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

RM 2,713.00


Estimated to ship on01 June 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer19)

Gene Information

human ... TH(7054)

General description

Th (tyrosine hydroxylase) is a homotetramer that has regulatory, catalytic and tetramerization domains. The gene is located on human chromosome llpl5.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tyrosine Hydroxylase around the phosphorylation site of Ser19.

Immunogen Range: 10-59

Biochem/physiol Actions

Th (tyrosine hydroxylase) plays an important role in catecholamine synthesis as it helps in the hydroxylation of L-tyrosine to l-DOPA (l-3,4-dihydroxyphenylalanine). Th gene mutation results in tyrosine hydroxylase deficiency.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brain catecholamine depletion and motor impairment in a Th knock-in mouse with type B tyrosine hydroxylase deficiency
Korner G, et al.
Brain (2015)
The rhombotin family of cysteine-rich LIM-domain oncogenes: distinct members are involved in T-cell translocations to human chromosomes 11p15 and 11p13
Boehm T, et al.
Proceedings of the National Academy of Sciences of the USA, 88(10), 4367-4371 (1991)
Izel Tekin et al.
Parkinsonism & related disorders, 23, 86-90 (2016-01-07)
Many of the symptoms and signs of Parkinson's disease (PD) arise from the death of midbrain dopamine neurons that utilize tyrosine hydroxylase (TH) as the rate-limiting enzyme in catecholamine biosynthesis. We investigated whether the presence of a common TH polymorphism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service