Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101794

Sigma-Aldrich

Anti-PHF6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BORJ, Anti-MGC14797, Anti-PHD finger protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
RM 2,107.00

RM 2,107.00


Estimated to ship on17 April 2025



Select a Size

Change View
100 μL
RM 2,107.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

RM 2,107.00


Estimated to ship on17 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PHF6(84295)

Immunogen

Synthetic peptide directed towards the C terminal region of human PHF6

Biochem/physiol Actions

PHF6 is a member of the plant homeodomain (PHD)-like finger (PHF) family. PHF6 is a protein with two PHD-type zinc finger domains, indicating a potential role in transcriptional regulation, that localizes to the nucleolus. Mutations affecting the coding region of its gene or the splicing of the transcript have been associated with Borjeson-Forssman-Lehmann syndrome (BFLS).This gene is a member of the plant homeodomain (PHD)-like finger (PHF) family. It encodes a protein with two PHD-type zinc finger domains, indicating a potential role in transcriptional regulation, that localizes to the nucleolus. Mutations affecting the coding region of this gene or the splicing of the transcript have been associated with Borjeson-Forssman-Lehmann syndrome (BFLS), a disorder characterized by mental retardation, epilepsy, hypogonadism, hypometabolism, obesity, swelling of subcutaneous tissue of the face, narrow palpebral fissures, and large ears. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Sequence

Synthetic peptide located within the following region: DFDIKTVLQEIKRGKRMVCSFYICYATLHLICCFKFRVHPKFIQSSENLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service