SAB1300001
Monoclonal Anti-Phospho-PLM-S68 antibody produced in mouse
purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-Phospholemman, FXYD1
Select a Size
RM 2,009.00
Estimated to ship on30 April 2025
Select a Size
About This Item
RM 2,009.00
Estimated to ship on30 April 2025
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
41AT858.235, monoclonal
form
buffered aqueous solution
species reactivity
mouse
technique(s)
indirect ELISA: 1:1000
western blot: 1:100-1:500
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pSer68)
Gene Information
mouse ... Fxyd1(56188)
Related Categories
General description
Immunogen
This monoclonal antibody is generated from mice immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S68 of mouse PLM.
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service