Skip to Content
Merck
All Photos(1)

Key Documents

N1773

Sigma-Aldrich

[Ser2]-Neuromedin C

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H72N16O11S
CAS Number:
Molecular Weight:
1093.26
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

[Ser2]-Neuromedin C, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

solid

composition

Peptide content, ~70%

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CC(C)C)NC(=O)C(Cc1cnc[nH]1)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc2c[nH]c3ccccc23)NC(=O)C(Cc4cnc[nH]4)NC(=O)C(CO)NC(=O)CN)C(C)C)C(N)=O

InChI

1S/C49H72N16O11S/c1-25(2)13-34(45(72)61-33(42(51)69)11-12-77-6)62-46(73)36(15-29-19-52-23-56-29)59-40(68)21-55-49(76)41(26(3)4)65-43(70)27(5)58-44(71)35(14-28-18-54-32-10-8-7-9-31(28)32)63-47(74)37(16-30-20-53-24-57-30)64-48(75)38(22-66)60-39(67)17-50/h7-10,18-20,23-27,33-38,41,54,66H,11-17,21-22,50H2,1-6H3,(H2,51,69)(H,52,56)(H,53,57)(H,55,76)(H,58,71)(H,59,68)(H,60,67)(H,61,72)(H,62,73)(H,63,74)(H,64,75)(H,65,70)

InChI key

CDSKUMVQOSVSRF-UHFFFAOYSA-N

Gene Information

human ... GRP(2922)

Amino Acid Sequence

Gly-Ser-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2

Biochem/physiol Actions

[Ser2]-Neuromedin C is a bombesin-like peptide known to inhibit feeding.

Other Notes

Bombesin-like peptide originally isolated from the brain of the European green frog, Rana ridibunda.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Mónika Fekete et al.
Brain research bulletin, 71(4), 386-392 (2007-01-09)
Bombesin-like peptides including gastrin releasing peptide and neuromedin C are known to inhibit feeding. Bombesin receptors have been found in moderate to high densities in the amygdaloid body, which is essentially involved in the regulation of feeding and body weight.
Biochemical and Biophysical Research Communications, 178, 529-529 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service